Would you like to log in again?
Human Recombinant 6-Phosphogluconate Dehydrogenase (from Cells)
Supplier: ProSci
92-379
75791-344EA 464.46 USD InStock
75791-344
Human Recombinant 6-Phosphogluconate Dehydrogenase (from Cells)
Enzymes
6-phosphogluconate dehydrogenase(PGD) is a cytoplasm-located protein, and belongs to the 6-phosphogluconate dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
6PGD is the second dehydrogenase in the pentose phosphate shunt. It catalyzes the oxidative decarboxylation of 6-phosphogluconate to ribulose 5-phosphate and CO2, with concomitant reduction of NADP to NADPH. Mutations within the gene coding this enzyme result in 6-phosphogluconate dehydrogenase deficiency, an autosomal hereditary disease effecting the red blood cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty