Would you like to log in again?
Human Recombinant Sedoheptulokinase (from Cells)
Supplier: ProSci
92-395
75791-360EA 946.47 USD InStock
75791-360
Human Recombinant Sedoheptulokinase (from Cells)
Enzymes
Sedoheptulokinase (SHPK) belongs to the FGGY kinase family, and is mainly located in cytoplasm
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SHPK is strongly expressed in liver, kidney and pancreas. It is expressed at lower levels in placenta and heart, and very weakly expressed in lung and brain. SHPK catalyzes the chemical reaction: ATP + sedoheptulose = ADP + sedoheptulose 7-phosphatecan, It can transform sedoheptulose to sedoheptulose 7-phosphate in the condition of ATP, and acts as a modulator of macrophage activation through control of glucose metabolism. In addition, It also can be down-regulated by LPS.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty