Would you like to log in again?
Mouse Recombinant 5'-Nucleotidase
Supplier: ProSci
92-401
75791-372EA 946.47 USD InStock
75791-372
Mouse Recombinant 5'-Nucleotidase
Enzymes
Mouse CD73 is a glycosyl phosphatidylinositol (GPI) anchored membrane protein that belongs to the 5'-nucleotidase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CD73 is an ecto 5'Nucleotidase expressed by most cell types. CD73 hydrolyzes extracellular nucleotides into membrane permeable nucleosides. CD73 is one of several enzymes responsible for the production of extracellular adenosine, a signaling molecule that is involved in responses to inflammation and tissue injury. CD73 is a lymphocyte maturation marker that has functions independent of its catalytic activity. CD73 is also a regulator of leukocyte extravasation, a function that requires its 5'Nucleotidase activity.CD73 has also been reported to regulate expression of pro-inflammatory molecules in mouse endothelium.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty