Would you like to log in again?
Mouse Recombinant Plasma Glutamate Carboxypeptidase
Supplier: ProSci
92-411
75791-392EA 946.47 USD InStock
75791-392
Mouse Recombinant Plasma Glutamate Carboxypeptidase
Enzymes
Carboxypeptidase Q (Cpq) is a member of the peptidase M28 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
PGCP is involved in a number of fundamental biological processes such as the hydrolysis of circulating peptides, catalyzing the hydrolysis of dipeptides with unsubstituted terminals into amino acids. Carboxypeptidase may play an important role in the liberation of thyroxine hormone from its thyroglobulin (Tg) precursor. The monomeric form is inactive while the homodimer is active.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty