Would you like to log in again?
Mouse Recombinant Carboxypeptidase A2
Supplier: ProSci
92-419
75791-408EA 744.04 USD InStock
75791-408
Mouse Recombinant Carboxypeptidase A2
Enzymes
Mouse carboxypeptidase A2(CPA2) is a secreted pancreatic procarboxy -peptidase which belongs to the peptidase M14 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
CPA2 consists of a signal peptide, a pro region and a mature chain. It can be activated after cleavage of the pro peptide. CPA2 cleaves the C-terminal amide or ester bond of peptides that have a free C-terminal carboxyl group. The hydrolytic action of CPA2 was identified with a preference towards long substrates with aromatic amino acids in their C-terminal end, particularly tryptophan.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty