Would you like to log in again?
Human Recombinant Aldehyde dehydrogenase 3-A1 (from Cells)
Supplier: ProSci
92-459
75791-488EA 743.82 USD InStock
75791-488
Human Recombinant Aldehyde dehydrogenase 3-A1 (from Cells)
Enzymes
Aldehyde dehydrogenase, dimeric NADP-preferring is an enzyme that in humans is encoded by the ALDH3A1 gene, belongs to the aldehyde dehydrogenase family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
ALDHs play a major role in the detoxification of alcohol-derived acetaldehyde. They are involved in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. This protein preferentially oxidizes aromatic aldehyde substrates. It may play a role in the oxidation of toxic aldehydes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty