Would you like to log in again?
Human Recombinant Biliverdin reductase A (from E. coli)
Supplier: ProSci
92-501
75791-572EA 744.04 USD InStock
75791-572
Human Recombinant Biliverdin reductase A (from E. coli)
Enzymes
Human Biliverdin reductase A (BLVRA) is belonged to the Gfo/Idh/MocA family and Biliverdin reductase subfamily
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
BLVRA is an enzyme that in humans is encoded by the BLVRA gene. BLVRA plays an important role in reducing the gamma-methene bridge of the open tetrapyrrole, biliverdin IX alpha, to bilirubin with the concomitant oxidation of a NADH or NADPH cofactor. BLVRA acts on biliverdin by reducing its double-bond between the pyrrole rings into a single-bond. It accomplishes this using NADPH + H+ as an electron donor, forming bilirubin and NADP+ as products.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty