Would you like to log in again?
S. hygroscopicus Recombinant PPT N-acetyltransferase (from E. coli)
Supplier: ProSci
92-657
75791-876EA 744.04 USD InStock
75791-876
S. hygroscopicus Recombinant PPT N-acetyltransferase (from E. coli)
Enzymes
Phosphinothricin N-acetyltransferase (PAT) is an enzyme that acetylates the free NH2 group of L-phosphinothricin (L-PPT) in the presence of acetyl-CoA as a co-substrate
This recombinant protein can be used for biological assays. For research use only.
It is highly specific for L-PPT and does not acetylate other L-amino acids or structurally similar molecules. L-PPT is a glutamate analog that can inhibit glutamine synthetase activity in plants, resulting in the accumulation of ammonia to toxic levels and impairment of photosynthesis. The introduction of a PAT gene into a plant genome can confer resistance to glufosinate herbicide during post-emergent applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty