Would you like to log in again?
K. pneumoniae Recombinant Kanamycin kinase type II (from E. coli)
Supplier: ProSci
92-658
75791-878EA 744.04 USD InStock
75791-878
K. pneumoniae Recombinant Kanamycin kinase type II (from E. coli)
Enzymes
Aminoglycoside 3'-phosphotransferase (APH(3')), also known as aminoglycoside kinase, is an aminoglycoside-modifying enzyme and widely presented in resistant bacteria
This recombinant protein can be used for biological assays. For research use only.
These ATP-dependent enzymes phosphorylate the 3'-hydroxyl of a variety of aminoglycosides including kanamycins, neomycins, paromomycins, neamine, ribostamycin, geneticin, and paromamine. These phosphorylated aminoglycosides fail to bind to their respective ribosomal binding sites with high affinity; hence resistance is conferred to the drugs that are phosphorylated. APH(3') is primarily found in certain species of gram-positive bacteria.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty