Would you like to log in again?
Human Recombinant IFN alpha 2b (from E. coli)
Supplier: ProSci
91-004
75791-996EA 464.32 USD InStock
75791-996
Human Recombinant IFN alpha 2b (from E. coli)
Proteins and Peptides
At least 23 different variants of IFN- alpha are known. The individual proteins have molecular masses between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. All IFN- alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN- alpha subtypes differ in their sequences by only one or two positions. Naturally occurring variants also include proteins that are truncated by 10 amino acids at the carboxyl-terminal end.
This recombinant protein can be used for biological assays. For research use only.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty