Would you like to log in again?
TUBE ERGOCLIP 8/12MM F/WEIGHING WITH XP
Supplier: Mettler Toledo
ErgoClip* Tube for Weighing^ ErgoClip* Tube for Weighing, use with XP, XS, XA analytical and XP, XS micro balance, Diameter: 8 / 12mm, Weighing device For PCR tubes, Eppendorf tubes, reaction tubes
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE ERGOCLIP 8/12MM F/WEIGHING WITH XP 75792-570 11106784 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty