Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Mouse Monoclonal Antibody [clone: AG103]

Supplier: ProSci
36-093
75792-854EA 589.13 USD InStock
75792-854
Anti-FTO Mouse Monoclonal Antibody [clone: AG103]
Antibodies
FTO (Fat mass-and obesity-associated gene) is the responsible gene for mouse ‘fused toes’ mutation. An association between FTO genotype and type 2 diabetes has been confirmed. The presence of the FTO rs9939609 A-allele was found to be positively correlated with other symptoms of the metabolic syndrome, including higher fasting insulin, glucose, triglycerides, and lower HDL-cholesterol.

ELISA: (direct and indirect: 1:2,000-1:5,000). Immunoprecipitation: (5ug/ml per sample in a final reaction volume of 500-1,000ul). Immunohistochemistry: (5-10ug/ml: paraffin sections). Western Blot: (1:2,000-1:5,000 using ECL. Suggested blocking and dilution buffer is PBST containing 0.05% Tween 20 and 5% skim milk. Suggested incubation time is 1 hour at room temperature). Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: Fto
Clonality: Monoclonal
Clone: AG103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty