Would you like to log in again?
Human Recombinant NAD Kinase (highly active) (from E. coli)
Supplier: ProSci
90-353
75794-172EA 700.27 USD InStock
75794-172
Human Recombinant NAD Kinase (highly active) (from E. coli)
Enzymes
NAD kinase catalyzes the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions
This recombinant proteins is for research use only.
Fusion-Tag: His Tag
NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty