Would you like to log in again?
Human Recombinant NAPRTase (from E. coli)
Supplier: ProSci
90-363
75794-320EA 827.54 USD InStock
75794-320
Human Recombinant NAPRTase (from E. coli)
Enzymes
Human nicotinate phosphoribosyltransferase (NAPRTase) is localized in the cytoplasm and is involved in biological processes such as NAD biosynthetic and metabolic processes, nicotinamide metabolic process, nicotinate nucleotide salvage, response to oxidative stress and water-soluble vitamin metabolic process
This recombinant proteins is for research use only.
Fusion-Tag: His Tag
It functions by catalyzing the conversion of nicotinic acid (NA) to NA mononucleotide (NaMN) and is essential for NA to increase cellular NAD levels and prevent oxidative stress of the cells. It is a crucial factor in the NAD+ biosynthesis pathway. Catalytic activity: Beta-nicotinate D-ribonucleotide + diphosphate = nicotinate + 5-phospho-alpha-D-ribose 1-diphosphate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty