Would you like to log in again?
Blue Light Conversion Screen
Supplier: Azure Biosystems
AC1025
75794-848EA 764.86 USD InStock
75794-848
Blue Light Conversion Screen
Photodocumentation Systems
Blue light conversion screen for transillumination of blue light.
  • Transillumination of blue light
  • Converts UV to blue light
  • Used for imaging in gel documentation systems
Blue light conversion screen for transillumination of blue light. Converts UV to blue light for imaging in gel documentation systems.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty