Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SLIDE CELL ANALYSIS ARTHUR 110X24X1.9 MM
Supplier: NANOENTEK USA INC.
Arthur* Cell analysis slide 50 slides/100 counts^ Arthur* Cell analysis slide, disposable, Contains Alignment beads, Material: Polymethy methacrylate (PMMA), Dimension: 110 mm (W) x 24 mm (D) x 1.9 mm (H), Chamber (sample) volume: 25 uL, for Image based cytometer, Size: 50 slides/100counts
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SLIDE CELL ANALYSIS ARTHUR 110X24X1.9 MM 75794-850 AC0050 Box of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty