Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ig Donkey Antibody (AzureSpectra 700)

Supplier: Azure Biosystems

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AC2156
75794-870EA 256.14 USD InStock
75794-870
Anti-Ig Donkey Antibody (AzureSpectra 700)
Antibodies
Azure offers a wide range of secondary antibodies labeled with fluorophores detected in infrared and near-infrared wavelengths.

AzureSpectra 700 and 800 secondary antibodies deliver enhanced sensitivity relative to visible fluorescent Western experiments because of low background autofluorescence in the near-infrared region.

Type: Secondary
Antigen Symbol: Ig
Clonality:
Clone:
Conjugation: AzureSpectra 700
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty