Would you like to log in again?
X20 Lightweight Proximity Jacket, Newtex Industries
Supplier: Newtex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2100394 2100395
75794-938EA 2625.56 USD InStock
75794-938 75794-936
X20 Lightweight Proximity Jacket, Newtex Industries
Workwear
X20 Lightweight Proximity Jackets are designed to protect against extreme radiant heat, sparks, and molten metal splash in environments with ambient temperatures up to 93 °C (200 °F).
  • Snap closures
  • Set-in sleeves with snaps on the cuffs for adjustable length
The Lightweight X20 Jacket has a two-layer construction, with a Z-Flex A-601 outer shell and an FR Cotton liner. Z-Flex reflects up to 95% of radiant heat.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty