Would you like to log in again?
Trident 287, Natural Rubber Gloves, 23" Mapa Professional
Supplier: Mapa Professionnel
287311 287310 287319
75794-972CS 863.96 USD InStock
75794-972 75794-968 75794-966
Trident 287, Natural Rubber Gloves, 23" Mapa Professional
Gloves
These gloves has high mechanical performance for long-lasting chemical protection.
  • Extra long length for added forearm protection
  • Provides protection against acids and alkalies/bases
  • Smooth chlorinated interior
Flexibility of natural rubber gives freedom of movement when worn over long periods of time.
Certifications: CE marked. EN 374: ABKL. EN 388: 2131.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty