Would you like to log in again?
Trident 286, 18" Natural Rubber Gloves, Mapa Professional
Supplier: Mapa Professionnel
286311 286310 286319
75795-034CS 756.6 USD InStock
75795-034 75794-970 75795-032
Trident 286, 18" Natural Rubber Gloves, Mapa Professional
Gloves
These natural rubber gloves has mechanical performance for long-lasting chemical protection.
  • Forearm protection
  • Chlorinated interior
  • Reinforced grip for better handling
Thickness of the material provides good impact protection, as well as a longer service life.
Certifications: CE Cat. III marked. EN 374: ABKL. EN 388: 2131
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty