Would you like to log in again?
GLOVE TEMP-DEX 720 PLU MAPA KNIT BK SZ9
Supplier: NEWELL BRANDS INC
Mapa* Temp-Dex 720 Plus Glove 9^ Mapa* Temp-Dex 720 Plus Glove, Material :Knit, Palm Coating: Nitrile, Temp-Dex 720, Lining: Terry Cloth, Finish/Grip on Glove Dotted, Cut Resistance CE, Level: 3, Color Black/Orange, Type of Cuff Knit: Wrist, Size : 9
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE TEMP-DEX 720 PLU MAPA KNIT BK SZ9 75795-030 720129ZQK Case of 72 Retrieving
Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty