Would you like to log in again?
Ozone Precursor/PAMS Mix, Restek
Supplier: Restek
26370
75795-048EA 5275 USD InStock
75795-048
Ozone Precursor/PAMS Mix, Restek
Standards Environmental Analysis Standards
Environmental air sampling gas standards meet lab requirements for two separate sources of calibration standards.
  • High-quality
  • No rental or demurrage fees required
  • Fast delivery provides freshest standard possible
Our high-quality air sampling gas calibration standards are provided by Spectra/Linde and Scott/Air Liquide—meeting lab requirements for two separate sources of calibration standards. Each comes with a certificate of analysis and unique serial number. All cylinders are disposable and do not require rental or demurrage fees. Recertification of cylinders is available directly with Restek's suppliers. All cylinders are drop-shipped from Restek's suppliers to provide fast delivery and the "freshest" standard possible. Minimum 12-month stability on all cylinders.
Ordering information: Ozone Precursor/PAMS Mix contains the following components at EPA concentrations: Acetylene, 40 ppbC ; Benzene, 30 ppbC; n-Butane, 40 ppbC; 1-Butene, 30 ppbC; cis-2-Butene, 35 ppbC; trans-2-Butene, 25 ppbC; Cyclohexane, 40 ppbC; Cyclopentane, 20 ppbC; n-Decane, 30 ppbC; m-Diethylbenzene, 40 ppbC; p-Diethylbenzene, 25 ppbC; 2,2-Dimethylbutane, 40 ppbC; 2,3-Dimethylbutane, 50 ppbC; 2,3-Dimethylpentane, 50 ppbC; 2,4-Dimethylpentane, 40 ppbC; n-Dodecane, 40 ppbC; Ethane, 25 ppbC; Ethylbenzene, 25 ppbC; Ethylene, 20 ppbC; m-Ethyltoluene, 25 ppbC; o-Ethyltoluene, 30 ppbC; p-Ethyltoluene, 40 ppbC; n-Heptane, 25 ppbC; n-Hexane, 30 ppbC; 1-Hexene, 60 ppbC; Isobutane, 25 ppbC; Isopentane, 40 ppbC; Isoprene, 40 ppbC; Isopropylbenzene, 40 ppbC; Methylcyclohexane, 30 ppbC; Methylcyclopentane, 25 ppbC; 2-Methylheptane, 25 ppbC; 3-Methylheptane, 25 ppbC; 2-Methylhexane, 25 ppbC; 3-Methylhexane, 25 ppbC; 2-Methylpentane, 20 ppbC; 3-Methylpentane, 40 ppbC; n-Nonane, 25 ppbC; n-Octane, 30 ppbC; n-Pentane, 25 ppbC; 1-Pentene, 25 ppbC; cis-2-Pentene, 35 ppbC; trans-2-Pentene, 25 ppbC; Propane, 40 ppbC; n-Propylbenzene, 30 ppbC; Propylene, 25 ppbC; Styrene, 40 ppbC; Toluene, 40 ppbC; 1,2,3-Trimethylbenzene, 25 ppbC; 1,2,4-Trimethylbenzene, 40 ppbC; 1,3,5-Trimethylbenzene, 25 ppbC; 2,2,4-Trimethylpentane, 30 ppbC; 2,3,4-Trimethylpentane, 25 ppbC; n-Undecane, 30 ppbC; o-Xylene, 25 ppbC; m/p-Xylene (combined), 40 ppbC.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty