Would you like to log in again?
Scott/Air Liquide Transportable Pure Gases and Mixtures, Restek
Supplier: Restek
34499 34521 34460 34524 34508 34478 34528 34483 34457 34543 34532 34472 34455 34515 34488 34474 34491 34514 34494 34453 34466 34461 34525 34531 34498 34459 34467 34451 34529 34519 34530 34522 34465 34469 34495 34493 34527 34468 34473 34492 34500 34505 34502 34496 34458 34537 34482 34485 34484 34449 34504 34501 34507 34476 34518 34480 34516 34479 34511 34463 34539 34534 34489 34452 34477 34462 34448 34487 34471 34497 34454 34464
75794-974EA 651.84 USD InStock
75794-974 75795-148 75795-106 75795-226 75795-188 75795-144 75795-066 75795-184 75795-140 10863-228 75795-062 75795-142 75795-064 75795-180 75795-182 75795-108 75795-228 75795-114 75795-158 75795-116 75795-078 75795-232 75795-110 75795-198 75795-156 75795-074 75795-194 75795-150 75795-076 75795-230 75795-152 75795-196 75795-072 75795-192 75794-992 75794-990 75795-118 75795-202 75795-004 75795-006 75795-204 75795-166 75795-088 75795-044 75795-000 75795-200 75795-162 75795-164 75795-080 75795-160 75795-082 75794-988 75794-986 76170-704 75795-090 76170-706 76170-708 75794-980 75795-206 75795-208 76170-702 75795-136 75795-138 75795-210 75795-132 75795-176 75795-014 75795-134 75795-178 75795-010 75795-092 75794-978
Scott/Air Liquide Transportable Pure Gases and Mixtures, Restek
Standards Environmental Analysis Standards
Transportable pure gases and mixtures are ideal for peak identification work.
  • More precise integration with analytical systems
  • Deliver large volumes of sample
  • Peak identification work
We offer a wide range of Scott/Air Liquide transportable gases, from pure gases for purging or calibrating to multicomponent mixes, which are ideal for peak identification work.The 14 L container has a CGA-160 connection for more precise integration with analytical systems. The 48 L cylinder has a CGA-165 connection and can deliver large volumes of sample. The 110 L cylinder has a CGA-180 connection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty