Transportable pure gases and mixtures are ideal for peak identification work.
- More precise integration with analytical systems
- Deliver large volumes of sample
- Peak identification work
We offer a wide range of Scott/Air Liquide transportable gases, from pure gases for purging or calibrating to multicomponent mixes, which are ideal for peak identification work.The 14 L container has a CGA-160 connection for more precise integration with analytical systems. The 48 L cylinder has a CGA-165 connection and can deliver large volumes of sample. The 110 L cylinder has a CGA-180 connection.