Would you like to log in again?
AlphaTec® 62-401 Chemical resistant gloves, Ansell
Supplier: Ansell
AlphaTec®
62401110 62401080 M9999821 113916 62401090
10017-172CS 541.3 USD InStock
10017-172 75795-324 10017-170 10017-176 10017-174
AlphaTec® 62-401 Chemical resistant gloves, Ansell
Gloves
Type 62-401 gloves have a double dipped natural rubber coating with wrinkle finish for added grip.
  • High degree of flexibility
  • Waterproof
  • Added grip
  • Thermal insulation in hot and cold extremes
These gloves are comfortable for periods of long use, offer a high level of flexibility, and are waterproof.
For use in construction, glass, public works, and food industries. Uses include handling sharp objects in a messy and/or humid environment.
62-401 gloves deliver thermal insulation in either hot (up to 250 °C/482 °F) or cold (down to −20 °C/−4 °F) conditions.
Certifications: 21 CFR (USA) compliant for food applications. ANSI Cut Level 2. ANSI Abrasion Level 3.
Packaging: Packaged 6 pairs per bag; 8 bags per carton.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty