Would you like to log in again?
AlphaTec® 23-201 Long-Sleeve PVC Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AlphaTec®
23201100 23201080 23201090
75795-562CS 1174.39 USD InStock
75795-562 75795-486 75795-560
AlphaTec® 23-201 Long-Sleeve PVC Gloves, Ansell
Gloves
Comfortable, long sleeve PVC glove, with good dexterity and pliability even at low temperatures.
  • Rough sandblast finish provides good wet grip
  • Medium-duty glove
  • Knitted cotton liner, 13 guage
  • Food contact approved
The AlphaTec® 23-201 glove features a soft PVC coating which maintains dexterity and pliability, even at low temperatures. It's dipped rough finish makes handling wet and slippery materials easier and safer. It is also heat sealed to offer liquid resistance and dust protection.
Certifications: EN 374; EN 420:2003 + A1:2009; Category lll; REACH compliant.
Packaging: Gloves are packaged 12 pairs per bag, 72 pairs per carton.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty