Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VAI® WipeDown HC Dry Wipes, Veltek Associates

Supplier: Veltek

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VAI®
VEL13-9X9-NS-HC-3024 VEL13-9X9-S-3013
75795-610CS 118.47 USD InStock
75795-610 76308-436
VAI® WipeDown HC Dry Wipes, Veltek Associates
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
WipeDown HC dry wipers are ultra-clean laundered and suitable for use in cleanroom environments.

  • Individually labeled with lot number and expiration date
  • Completely lot traceable
  • High absorbency and low in particulate shedding and soluble extracts
  • Validated for sterility and shelf life

Constructed of 55% polyester and 45% cellulose spunlace blend. Particulates, residues, and foreign particles are effectively entrapped in the webbing of both of these materials.

Uses a proprietary cutting method, FocusEdge heat cutting technology. This ensures that there is a reduction in edge particulates on the wiper that is typically left behind after laser cutting.

WipeDown HC dry wipers can be used for filling and packaging machinery wipe downs, IPA wipe downs, spill control, general wiping, and contamination control.

Certifications: Sterility tested according to the USP compendium with lot specific certificates of irradiation, certificates of sterility, and certificates of conformance available upon delivery. Sterile versions are sterility assured through gamma irradiation at a 10⁻⁶ SAL.

Delivery information: Delivered with lot specific documentation.

Packaging: Packaged neatly folded for easy removal.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty