Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VAI® ALCOH-WIPE® Wipes, Veltek Associates

Supplier: Veltek

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VAI®
VEL6-6X6-S-2307 VEL6-18X18-S-2304 VEL6-12X12-S-2302
75795-624CS 672 USD InStock
75795-624 89173-568 75795-626
VAI® ALCOH-WIPE® Wipes, Veltek Associates
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Wipes saturated with 70% USP isopropyl alcohol and 30% USP water for injection.

  • Validated for sterility and shelf life
  • 100% synthetic filament constructed in a continuous knit that minimizes fiber and particulate release

Use to wipe down filling and packing machinery and for spill control on lexan, glass, polycarbonate, and stainless steel.

Ordering information: Available as individual packs, 100 per case.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty