Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Round Carboy with Sanitary Neck
Supplier: VWR
VWR®
584301 584303 584302
75795-678CS 1369.63 USD InStock
75795-678 75795-684 75795-692
VWR® Round Carboy with Sanitary Neck
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for bioprocessing applications.
  • Non sterile
  • Polypropylene formats available
  • Excellent chemical resistant
  • Narrow-mouth
  • Feature an integrated shoulder hand
  • Features a 76 mm (3") sanitary flange that accepts standard TC fittings
Carboys are well suited for use in temperatures ranging from -100 to 100 °C (-148 to 212 °F). Carboys are leak-proof under standard test conditions.
Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth.
Certifications: Resin meets US FDA 21 CFR, USP Class VI.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty