Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Round Carboy with Sanitary Flange, Narrow Mouth

Supplier: VWR
VWR®
584306 584305
75795-686EA 681.08 USD InStock
75795-686 75795-694
VWR® Round Carboy with Sanitary Flange, Narrow Mouth
Bottles Carboys
Use for storing and dispensing solutions and media. Ideal for sterile water storage.

  • Non sterile, excellent chemical resistance
  • Polypropylene formats available
  • Clear graduation marks
  • Narrow-mouth
  • Feature an integrated shoulder handle
  • Feature a 41 mm sanitary flange for use as a dispensing port

Carboys are well suited for use in temperatures ranging from –100 to 100 °C (–148 to 212 °F). Carboys are leak-proof under standard test conditions.

Polypropylene carboys can be autoclaved before refilling to stop recurring bacterial growth. Provides a secure hookup for use as a supply reservoir.

Certifications: Resin meets US FDA 21 CFR, USP Class VI.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty