Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Kimax® Petri Dish Sets, Kimble Chase
Supplier: Kimble Chase
KIMAX®
23061-10015 23061-6015 23061-10020
75996-750CS 323.84 USD InStock
75996-750 75795-713 75795-715
Kimax® Petri Dish Sets, Kimble Chase
Culture Dishes Petri Dishes
Cell culture dishes feature reinforced beaded edges to resist mechanical breakage and help center the dish cover
These petri dish sets are made up of glass.
Dishes remain clear even after repeated use in wet or dry sterilization cycles. Covers have a fused-on bright red ceramic enamel marking spot and the inscription “COVER.” Bottoms have a white ceramic enamel inscription that reads “BOTTOM.” The combination of different colors and wording permits quick and easy identification of the parts when sorting, selecting, and assembling pairs. Bottoms also have a vertical arrow on the side to precisely locate serial dilution starting points. Complete dish sets include covers and bottoms, packed in separate boxes. Designed from Federal Specification NNN-D-1478.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty