Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Support Stand and Clamps for Sonifier® Cell Disruptors, Branson Ultrasonics

Supplier: Branson

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

101-063-1110 101-063-1109 101-063-1108
75795-832EA 83.64 USD InStock
75795-832 75795-834 75795-836
Support Stand and Clamps for Sonifier® Cell Disruptors, Branson Ultrasonics
Support Stands
The support stand with stainless steel rod accommodates Branson’s Sonifier® line and accessories

  • Stand base measures 22.9L x 13.9W x 60.9H cm (9 x 5.5 x 24")
  • Stainless steel rod measures 60.9H x 1.3dia. cm (24 x 0.5")

The converter clamp supports the ultrasonic stack (converter and horn) and is easily adjusted to properly position the horn in the sample. The stand features a black enameled cast-iron base with zinc-plated rod.

Ordering information: Support stand with Rod is supplied without clamps, which must be purchased separately. Clamp orders include clamp only.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty