Would you like to log in again?
CELLCOAT® Protein Coated Flasks, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CELLCOAT®
658940 690940-010 658950-005 661940 690950-010 658940-005 690950 658950 661950 690940
82050-862CS 862.36 USD InStock
82050-862 82050-884 75795-994 82050-864 82050-886 75795-996 75796-006 82051-086 75796-004 82051-084
CELLCOAT® Protein Coated Flasks, Greiner Bio-One
Flasks Cell Culture Flasks
These convenient, ready-to-use tissue culture flasks are coated with extra-cellular matrix proteins (ECM) or synthetic peptides.
  • High-profile design
  • Nonpyrogenic
  • Free of RNase and DNase
  • Convenient and ready to use
Coating provides an inert scaffolding that helps regulate cell growth and proliferation. It simulates the cells' natural environment, which facilitates adherence, differentiation, proliferation, and longevity in many cell types. Flasks also feature a canted neck, plug seal, "Best Before" date, and a lot number on each pack. Flasks are nonpyrogenic and DNase- and RNase-free. All 650 mL flasks feature a high profile design.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty