Would you like to log in again?
Neoprene Gloves, Mapa Professional
Supplier: Mapa Professionnel
34382040 34382048 34382047 34382041 34382049
75796-100CS 1135.01 USD InStock
75796-100 75796-098 75796-092 75796-096 75796-094
Neoprene Gloves, Mapa Professional
Gloves
Chemical resistant neoprene gloves with pinked cuff type and Z pattern finish.
  • Embossed texture for better grip on wet objects
  • Added comfort and thermal insulation, due to the cotton-knit lining
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty