Would you like to log in again?
Sapphire PCR Tubes and Strips, Polypropylene, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
373274 684201 373281 673281 671277 683271 671276 373270 673210 682276 673282 683274 683281 373273 683276 373275 671231 673287 683275 682273 683277 673285 682277 683201 671281 373293 673280 671273 673274 373292 671274 373277 673286 673273 608201 673277 673275 673271 671221 682201 373276 373250 671275 683273 682274 671241 673276 673284 608281 682281 682275 671201 673283
75796-144CS 1377 USD InStock
75796-144 75820-360 75796-142 75796-148 75786-259 75796-146 75796-140 75830-576 75830-574 10859-842 75796-156 75796-154 75796-158 75796-152 75830-558 75796-150 75830-556 75786-257 75796-108 75796-166 75796-122 75796-120 75796-164 75796-126 75796-124 75796-168 75796-162 75796-160 75816-118 89204-786 75830-554 75830-552 75796-118 75796-178 75796-134 75831-386 75796-176 75796-132 75831-388 75796-138 75796-136 75796-170 75796-174 75796-130 75796-172 75830-542 75816-128 75830-540 75816-122 75816-120 75796-128 75816-126 75816-124
Sapphire PCR Tubes and Strips, Polypropylene, Greiner Bio-One
Tubes PCR Tubes
Sapphire PCR Tubes and strips feature attached or unattached caps in either domed or flat designs, and frosted caps are available for easy labeling.
  • Manufactured out of medical-grade polypropylene without interfering additives
  • Thin-walled for efficient heat transfer during amplification
  • Options for real-time/quantitative PCR
  • Comprehensive thermal cycler compatibility
  • Packaging labeled with product symbols for easier working and warehousing
Maximum volume capacity ranges between models. Tube strips are available in formats of standard or low profile.
Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty