Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SafeCup™ lll MultiDrug Panel, Germaine® Laboratories
Supplier: Germaine Laboratories
SafeCup™ III
45485 40105 45475 41577 40675
75796-236CS 257.67 USD InStock
75796-236 75796-216 75796-238 75799-284 75796-182
SafeCup™ lll MultiDrug Panel, Germaine® Laboratories
Drug Tests
SafeCup™ lll Round Cups are a series of All-in-One (collection/testing/transport) qualitative tests for the detection of drugs of abuse in urine
Tests include 25 collection cups, security seals, gloves, and instructions.
Tests adhere to SAMSHA cutoff levels. Cups feature a leak-proof cap, temperature strip, and security seal included.
Certifications: CLIA Waived.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty