Would you like to log in again?
SpectraMax® Quant™ dsDNA Assay Kits, Molecular Devices
Supplier: Molecular Devices
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
R8354 R8359 R8356 R8358 R8357 R8355 R8359 R8354 R8358 R8356 R8357 R8355
75796-320EA 477.93 USD InStock
75796-320 CA75796-322 75796-324 CA75796-320 75796-322 75796-318 CA75796-316 CA75796-326 75796-316 75796-326 CA75796-324 CA75796-318
SpectraMax® Quant™ dsDNA Assay Kits, Molecular Devices
Assays Cellular Assays
The SpectraMax® Quant™ dsDNA Assay Kits are designed for fluorescence-based dsDNA quantitation across a broad range of concentrations.
  • Tailored to your different needs
  • Configured and optimized for microplate readers
  • Sensitive, accurate, and easy to use
The SpectraMax® Quant™ dsDNA Assay Kits are designed for fluorescence-based dsDNA quantitation across a broad range of concentrations
Quantitation of DNA is a critical step in molecular biology requiring accuracy, reliability, and the use of increasingly smaller sample volumes for applications such as next-generation sequencing. Compared to spectrophotometric DNA quantification, the fluorometric method provides key advantages such as significantly increased sensitivity, high selectivity for double-stranded DNA (dsDNA) over single-stranded DNA (ssDNA) or RNA, and improved contaminant tolerance (protein and carbohydrate molecules). The SpectraMax Quant dsDNA Assay Kits provide pre-optimized, complete solutions that are sensitive, accurate, and easy to use.
Tailored to your different needs, these assay kits are configured and optimized for Molecular Devices SpectraMax® microplate readers, with preconfigured protocols provided in SoftMax® Pro Software for simplified data acquisition and analysis.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty