Would you like to log in again?
AimStrip® Capillary Transfer Tubes, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip®
78155
75796-386BX 37.92 USD InStock
75796-386
AimStrip® Capillary Transfer Tubes, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Plastic capillary tubes are designed to collect blood specimens up to 10µL
Tubes feature a built-in air vent for easy dispensing of specimen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty