Would you like to log in again?
Repeater® E3 and E3x Pipettors, Eppendorf®
Supplier: Eppendorf
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Repeater®
4987000398 4987000398 4987000410 4987000118 4987000134 4987000410
75796-402EA 2155.3 USD InStock
75796-402 75796-406 CA75796-402 75796-404 CA75796-408 75796-408
Repeater® E3 and E3x Pipettors, Eppendorf®
Pipettes Repetitive Pipettors
The Repeater® E3/E3x Pipettors, in combination with the Combitips Advanced® pipette tips, creates a versatile dispensing system that functions according to the positive displacement principle.
  • Automatic Combitip® recognition
  • Ergonomic one-button tip ejector
  • Precisely programmable aspiration and dispensing speeds
  • Repetitive dispensing modes
  • Wide dispensing range: 1 μl to 50 ml, in 100-nanoliter increments
  • Safe and ergonomic operation with 9 different selectable languages
These devices provide precise and accurate dispensing of viscous solutions (e.g. glycerol) or liquids with high vapor pressure (e.g. ethanol). Contamination-free dispensing for toxic, radioactive, and other hazardous liquids (all available modes). Large illuminated color display reduces eye strain when reading. Motor-driven function reduces human error to a minimum, and reduces the risk of repetitive strain injuries (RSI).
It is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty