Would you like to log in again?
CHAMBER SFTY DRYING FDL115 230V 5-300 C
Supplier: BINDER INC
Safety drying chambers, Model FDL 115^ Safety drying chambers, Model FDL 115, Standard, 230 V 1 approx 50/60 Hz, Temperature range: room temperature plus 5 deg C to 300 deg C, Replaceable filter cartridge, Class F6, 2 chrome-plated racks, Audible and visual alarm and automatic
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHAMBER SFTY DRYING FDL115 230V 5-300 C 75796-632 9010-0292 Each (115l) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty