Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum Crucibles, Heraeus

Supplier: Heraeus

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TI1/5 TI3/11 TI1/12 TI1/8 TI1/10 TI3/12 TI3/3 TI1/4 TI1/11 TI3/8 TI3/9 TI1/1 TI3/4 TI1/3 TI3/1 TI1/2 TI1/6 TI3/6 TI3/5 TI3/7 TI1/9 TI3/10 TI1/7 TI3/2
75796-992EA 4003.71 USD InStock
75796-992 75796-970 75796-980 75796-990 75796-974 75796-996 75796-962 75796-984 75796-940 75796-972 75796-994 75796-982 75796-960 75797-000 75796-978 75796-988 75796-966 75796-998 75796-976 75796-942 75796-964 75796-986 75796-938 75796-968
Platinum Crucibles, Heraeus
Crucibles
These Crucibles are made with 99.95% Pure Platinum

Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Crucibles have become essential for the preparation of samples in analytical laboratories.

They are available in Standard Form or Wide Form.

Ordering information: Lids sold separately. For additional lid details, see Lids for Platinum and Platinum 95%/Gold 5% Crucibles, Heraeus.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty