Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum 95%/Gold 5% Crucibles, Heraeus

Supplier: Heraeus

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

TI3/9-PTAU5 TI1/5-PTAU5 TI3/4-PTAU5 TI3/1-PTAU5 TI3/2-PTAU5 TI3/11-PTAU5 TI1/12-PTAU5 TI3/6-PTAU5 TI1/2-PTAU5 TI1/1-PTAU5 TI1/10-PTAU5 TI3/10-PTAU5 TI1/8-PTAU5 TI3/5-PTAU5 TI1/6-PTAU5 TI1/7-PTAU5 TI1/11-PTAU5 TI3/3-PTAU5 TI1/4-PTAU5 TI1/3-PTAU5 TI3/7-PTAU5 TI3/12-PTAU5 TI1/9-PTAU5 TI3/8-PTAU5
75797-018EA 4857.84 USD InStock
75797-018 75797-028 75797-006 75797-008 75797-010 75797-032 75797-042 75797-020 75797-034 75797-012 75797-044 75797-022 75797-036 75797-014 75797-046 75797-024 75797-002 75797-016 75797-038 75797-026 75797-004 75797-048 75797-030 75797-040
Platinum 95%/Gold 5% Crucibles, Heraeus
Crucibles
These crucibles are made with 95% Platinum and 5% Gold

Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Crucibles have become essential for the preparation of samples in analytical laboratories.

The mechanical stability of Platinum is further improved with the addition of 5% Gold. This alloy offers higher strength and low wettability, so that the sample can be easily removed from the crucible.

They are available in Standard Form or Wide Form.

Ordering information: Lids sold separately.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty