Would you like to log in again?
COLUMN C8 ALLTIMA HP 5UM REVRS 100X1.0MM
Supplier: HICHROM LIMITED
Alltima* HP C8 Column 100x1.0mm^ Alltima* HP C8 Column, Particle size: 5um, Chromatographic Properties: Lower capacity compared to C18 phases, Application: Reversed-phase applications where C18 is too retentive, Dimension: 100x1.0mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN C8 ALLTIMA HP 5UM REVRS 100X1.0MM 75797-582 22473 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty