Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BootieButler® Shoe Covers, Protexer
Supplier: Protexer
BootieButler®
KBNS40-600-ESD KBNS40-600-FAS KBNS40-600 KBNS28-750 KBNS28-750-FAS KBNS28-1-ESD KBNS28-1-FAS KBNS28-750-ESD
75796-908CS 1775.9 USD InStock
75796-908 75798-788 75798-786 10836-051 75798-784 76186-138 75798-782 75798-790
BootieButler® Shoe Covers, Protexer
Disposable Garments
The original BootieButler® Shoe Cover is an economical anti-skid and anti-static shoe cover that serves well in a variety of situations
  • Super anti-skid, slip-resistant pattern
  • Antistatic material
  • Lightweight and durable for extra-long wear
  • One size fits most
  • Specifically designed for use in the BootieButler® automatic dispenser
BootieButler® Shoe Covers are ideal for use in cleanrooms, food processing, real estate, healthcare, construction, manufacturing, research and development, and energy applications.
Constructed of durable spunbond polypropylene material with non-skid, non-conductive soles. The skid-resistant pattern improves friction and traction. A strong elastic band around the ankle allows for a universal fit.
Certifications: Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5.
Ordering information: These shoe covers are designed to be used in the BootieButler® automatic dispenser, and not as a bulk product.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty