Would you like to log in again?
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
28107PTD5/600 25-28107
75798-804BX 131.97 USD InStock
75798-804 75796-936
Puritan® MALDI-TOF Pointed Applicator Stick, Puritan Medical Products
Swabs Swabs for Sampling
This 73mm pointed applicator stick is designed for use as a colony transfer device with the automated MALDI-TOF MS systems
Made in the USA.
The stick is made out of Northern Maine White Birch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty