Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L1
Classic reversed phase retention and selectivity. High quality hydrophobic general purpose C18.