Would you like to log in again?
Superdex Columns, Peptide 75 Increase, Cytiva
Supplier: Cytiva
Superdex™
29148721 29-1487-21 29-1487-22 29148723 29-1487-23 29148722
75799-302EA 3229.6 USD InStock
CA75802-916 CA75802-902 75799-302 75799-304 75799-300 CA75802-918
Superdex Columns, Peptide 75 Increase, Cytiva
Chromatography Columns
Superdex 75 Increase 10/300 GL, 5/150 GL, and 3.2/300 are prepacked columns for versatile use in size exclusion chromatography applications.
  • Up to 50% higher resolution compared with predecessor Superdex 75, for improved purity and analysis results
  • Runtime reduced down to one third compared with predecessor Superdex 75, with maintained resolution for results faster
  • Versatile use in both preparative and analytical applications, especially with recombinant tagged proteins
  • Higher lot-to-lot consistency compared with predecessor Superdex 75
  • Tolerance to repeated harsh cleaning procedures at high pH ensures long column lifetime and minimizes carryover
The columns are intended for use in preparative purification as well as for characterization and analysis of proteins with molecular weights between Mr 3000 and 70 000, such as recombinant tagged proteins.
Superdex 75 Increase belongs to the new generation of agarose based SEC media, comprising smaller, more rigid beads with a narrower particle size distribution and a higher selectivity for improved performance compared with its predecessor Superdex 75.
Superdex 75 Increase resin is alkali-resistant. Hence, the column can be cleaned in place under alkali conditions to allow use of the same column for different proteins with minimal risk for carryover between samples. The alkali resistance is what differentiates agarose-based resins from silica-based resins. The possibility to clean the resin under harsh alkali conditions also ensures a long column lifespan.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty