Would you like to log in again?
Screw Caps for Internal Thread Sample Tubes, Simport Scientific
Supplier: Simport
T500OOS T500YOS T500GOS T500NOS T500ROS T500BOS T500WOS T500LOS
75799-290PK 178.34 USD InStock
75799-290 75799-316 75799-292 75799-298 75799-286 75799-288 75799-294 75799-296
Screw Caps for Internal Thread Sample Tubes, Simport Scientific
Closures Test/Sample Tube Closures
These PP screw caps are to be used with T500 series internal thread sample tubes.
  • Silicone washer seal
  • Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
Ordering information: Tubes T500 series sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty