Would you like to log in again?
CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner bio-one
CELLSTAR®
627160 639160 628160 628160-TRI 664160 639160-TRI 664160-TRI
82050-598CS 158.47 USD InStock
82050-598 75799-856 75799-858 82050-546 82050-538 75799-854 82050-916
CELLSTAR® Tissue Culture Dishes, Polystyrene, Sterile, Greiner Bio-One
Culture Dishes Cell Culture Dishes
CELLSTAR® dishes are manufactured from high-quality virgin polystyrene and are treated for optimal cell attachment and proliferation.
  • High-quality virgin polystyrene construction
  • Nonpyrogenic
  • Free of RNase and DNase
  • Stackable
  • Uniform gas exchange and reduced condensation
Dishes offer distortion-free clarity. They are nonpyrogenic and RNase- and DNase-free. All dishes feature stacking rims for simple storage and ease in handling. Vented versions provide uniform gas exchange and reduced condensation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty