Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Centrifuge Tubes
Supplier: TradeWinds (TWD)
10-9501 10-9152 BX10-0501 10-9502 BX10-0502 BX10-0151 10-9151 BX10-0152
75799-900CS 333.2 USD InStock
75799-900 75799-888 75799-896 75799-898 75799-886 75799-892 75799-894 75799-890
Centrifuge Tubes
Tubes Centrifuge Tubes
Centrifuge tubes feature conical bottoms for superior separation and pelleting.
  • Gamma radiation sterilized
  • Non-pyrogenic
  • 15 and 50 ml conical tubes
  • Autoclavable
  • Graduated
  • Tubes withstand centrifugation of 8400 RCF
  • Medical-grade polypropylene construction
  • Transparent wall permits easy viewing of tube content
Tubes are supplied with either a Plug-Seal cap or a Flat-Top cap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty